Få mere at vide
rac-N-Benzyl-N-[2-hydroxyl-2-(4-benzyloxy-3-nitrophenyl)-ethyl]-3-(4-methoxyphenyl)-2-propylamine-d6, TRC
Artikelnummer. 30341412
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30341412 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30341412 Leverandør Toronto Research Chemicals Leverandørnr. TRCB279102100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[benzyl-[1,1,1-trideuterio-3-[4-(trideuteriomethoxy)phenyl]propan-2-yl]amino]-1-(3-nitro-4-phenylmethoxyphenyl)ethanol
CAS 1216475-21-6
Molekylær formel C32 2H6 H28 N2 O5
Mængde 100 mg
InChI formel InChI=1S/C32H34N2O5/c1-24(19-25-13-16-29(38-2)17-14-25)33(21-26-9-5-3-6-10-26)22-31(35)28-15-18-32(30(20-28)34(36)37)39-23-27-11-7-4-8-12-27/h3-18,20,24,31,35H,19,21-23H2,1-2H3/i1D3,2D3
SMIL [2H]C([2H])([2H])Oc1ccc(CC(N(CC(O)c2ccc(OCc3ccccc3)c(c2)[N+](=O)[O-])Cc4ccccc4)C([2H])([2H])[2H])cc1
IUPAC navn 2-[benzyl-[1,1,1-trideuterio-3-[4-(trideuteriomethoxy)phenyl]propan-2-yl]amino]-1-(3-nitro-4-phenylmethoxyphenyl)ethanol
Molekylvægt (g/mol) 532.66
Formel vægt 532.2844