Få mere at vide
rac-N-Benzyl-N-[2-hydroxyl-2-(4-benzyloxy-3-formamidophenyl)-ethyl]-3-(4-methoxyphenyl)-2-propylamine-d6, TRC
Artikelnummer. 30343054
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30343054 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30343054 Leverandør Toronto Research Chemicals Leverandørnr. TRCB27909710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[5-[2-[benzyl-[1,1,1,2,3,3-hexadeuterio-3-(4-methoxyphenyl)propan-2-yl]amino]-1-hydroxyethyl]-2-phenylmethoxyphenyl]formamide
CAS 1216518-77-2
Molekylær formel C33 2H6 H30 N2 O4
Mængde 10 mg
InChI formel InChI=1S/C33H36N2O4/c1-25(19-26-13-16-30(38-2)17-14-26)35(21-27-9-5-3-6-10-27)22-32(37)29-15-18-33(31(20-29)34-24-36)39-23-28-11-7-4-8-12-28/h3-18,20,24-25,32,37H,19,21-23H2,1-2H3,(H,34,36)/i1D3,19D2,25D
SMIL [2H]C([2H])([2H])C([2H])(N(CC(O)c1ccc(OCc2ccccc2)c(NC=O)c1)Cc3ccccc3)C([2H])([2H])c4ccc(OC)cc4
IUPAC navn N-[5-[2-[benzyl-[1,1,1,2,3,3-hexadeuterio-3-(4-methoxyphenyl)propan-2-yl]amino]-1-hydroxyethyl]-2-phenylmethoxyphenyl]formamide
Molekylvægt (g/mol) 530.69
Formel vægt 530.3052