Få mere at vide
rac N-[4-Chloro-2-[3-cyclopropyl-1-hydroxy-1-(trifluoromethyl)-2-propynyl]phenyl]-4-methoxybenzamide, TRC
Artikelnummer. 30354957
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30354957 2.5 mg
30363735 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30354957 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3651502.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1189491-03-9
Molekylær formel C21H17ClF3NO3
Molekylvægt (g/mol) 423.81
Synonym SW 965
IUPAC navn N-[4-chloro-2-(4-cyclopropyl-1,1,1-trifluoro-2-hydroxybut-3-yn-2-yl)phenyl]-4-methoxybenzamide
SMIL COc1ccc(cc1)C(=O)Nc2ccc(Cl)cc2C(O)(C#CC3CC3)C(F)(F)F
Tekniske data
Kemisk navn eller materiale rac N-[4-Chloro-2-[3-cyclopropyl-1-hydroxy-1-(trifluoromethyl)-2-propynyl]phenyl]-4-methoxybenzamide
Mængde 2.5 mg
InChI formel InChI=1S/C21H17ClF3NO3/c1-29-16-7-4-14(5-8-16)19(27)26-18-9-6-15(22)12-17(18)20(28,21(23,24)25)11-10-13-2-3-13/h4-9,12-13,28H,2-3H2,1H3,(H,26,27)