Få mere at vide
rac Mono(5-carboxy-2-ethylpentyl) Phthalate, TRC
Artikelnummer. 17980395
Skift visning
Mængde:
1 mg
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17980395 2.5 mg 2.5mg
17954435 1 mg -
17965045 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17980395 Leverandør Toronto Research Chemicals Leverandørnr. TRCM5255502.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 40809-41-4
Molekylær formel C16 H20 O6
Molekylvægt (g/mol) 308.33
Synonym Phthalic acid, mono-5-carboxy-2-ethylpentyl ester
IUPAC navn 2-(5-carboxy-2-ethylpentoxy)carbonylbenzoic acid
SMIL CCC(CCCC(=O)O)COC(=O)c1ccccc1C(=O)O
Tekniske data
Kemisk navn eller materiale rac Mono(5-carboxy-2-ethylpentyl) Phthalate
Renhedsgrad noter HPLC
Mængde 2.5 mg
InChI formel InChI=1S/C16H20O6/c1-2-11(6-5-9-14(17)18)10-22-16(21)13-8-4-3-7-12(13)15(19)20/h3-4,7-8,11H,2,5-6,9-10H2,1H3,(H,17,18)(H,19,20)
Formel vægt 308.126
Procent renhed >95%