Få mere at vide
rac Mirabegron-d5, TRC
Artikelnummer. 30481356
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30481356 10 mg
30469739 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30481356 Leverandør Toronto Research Chemicals Leverandørnr. TRCM36490210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1215807-38-7
Molekylær formel C21H19D5N4O2S
Molekylvægt (g/mol) 401.54
Synonym 2-Amino-N-[4-[2-[[2-hydroxy-2-phenylethyl]amino]ethyl]phenyl]-4-thiazoleacetamide-d5, 2-(2-Aminothiazol-4-yl)-4'-[2-[(2-hydroxy-2-phenylethyl)amino]ethyl]acetic Acid Anilide-d5, YM 178-d5
SMIL NC1=NC(CC(NC2=CC=C(CCNCC(C3=C([2H])C([2H])=C([2H])C([2H])=C3[2H])O)C=C2)=O)=CS1
Tekniske data
Kemisk navn eller materiale rac Mirabegron-d5
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C21H24N4O2S/c22-21-25-18(14-28-21)12-20(27)24-17-8-6-15(7-9-17)10-11-23-13-19(26)16-4-2-1-3-5-16/h1-9,14,19,23,26H,10-13H2,(H2,22,25)(H,24,27)/i1D,2D,3D,4D,5D
Procent renhed >95%