Få mere at vide
rac Metoprolol-d7, TRC
Artikelnummer. 30476753
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30476753 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476753 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33879210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Metoprolol-d7
CAS 959787-96-3
Renhedsgrad noter HPLC
Molekylær formel C15 2H7 H18 N O3
Mængde 10 mg
Synonym (±)-Metoprolol-(isopropyl-d7), 2-Propanol, 1-[4-(2-methoxyethyl)phenoxy]-3-[[1-(methyl-d3)ethyl-1,2,2,2-d4]amino]-, Metoprolol D7 (isopropyl D7)
InChI formel InChI=1S/C15H25NO3/c1-12(2)16-10-14(17)11-19-15-6-4-13(5-7-15)8-9-18-3/h4-7,12,14,16-17H,8-11H2,1-3H3/i1D3,2D3,12D
SMIL [2H]C([2H])([2H])C([2H])(NCC(O)COc1ccc(CCOC)cc1)C([2H])([2H])[2H]
IUPAC navn 1-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)-3-[4-(2-methoxyethyl)phenoxy]propan-2-ol
Molekylvægt (g/mol) 274.41
Formel vægt 274.2274
Procent renhed >95