Få mere at vide
rac Mephenytoin-d5, TRC
Artikelnummer. 30471658
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30471658 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30471658 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2250071MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Mephenytoin-d5
CAS 1185032-66-9
Molekylær formel C12 2H5 H9 N2 O2
Mængde 1 mg
Synonym 5-Ethyl-3-methyl-5-(2,3,4,5,6-pentadeuteriophenyl)imidazolidine-2,4-dione, 5-Ethyl-3-methyl-5-(phenyl-d5)-2,4-imidazolidinedione, 5-Ethyl-3-methyl-5-(phenyl-d5)hydantoin, Mephenytoin-d5, Mephenytoin-D5 (phenyl-D5)
InChI formel InChI=1S/C12H14N2O2/c1-3-12(9-7-5-4-6-8-9)10(15)14(2)11(16)13-12/h4-8H,3H2,1-2H3,(H,13,16)/i4D,5D,6D,7D,8D
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C2(CC)NC(=O)N(C)C2=O
IUPAC navn 5-ethyl-3-methyl-5-(2,3,4,5,6-pentadeuteriophenyl)imidazolidine-2,4-dione
Molekylvægt (g/mol) 223.28
Formel vægt 223.1369