Få mere at vide
rac Mephenytoin-d3, TRC
Artikelnummer. 30468497
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30468497 2.5 mg 2.5mg
30465610 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30468497 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2250022.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1185101-86-3
Molekylær formel C12 2H3 H11 N2 O2
Molekylvægt (g/mol) 221.27
Synonym 5-Ethyl-5-phenyl-3-(trideuteriomethyl)imidazolidine-2,4-dione, 5-Ethyl-3-(methyl-d3)-5-phenyl-2,4-imidazolidinedione, 5-Ethyl-3-(methyl-d3)-5-phenylhydantoin, Mephenytoin-d3, Mephenytoin-D3 (N-methyl-D3)
IUPAC navn 5-ethyl-5-phenyl-3-(trideuteriomethyl)imidazolidine-2,4-dione
SMIL [2H]C([2H])([2H])N1C(=O)NC(CC)(C1=O)c2ccccc2
Tekniske data
Kemisk navn eller materiale rac Mephenytoin-d3
Mængde 2.5 mg
InChI formel InChI=1S/C12H14N2O2/c1-3-12(9-7-5-4-6-8-9)10(15)14(2)11(16)13-12/h4-8H,3H2,1-2H3,(H,13,16)/i2D3
Formel vægt 221.1244