Få mere at vide
rac Matairesinol-d6, TRC
Artikelnummer. 30473800
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30473800 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473800 Leverandør Toronto Research Chemicals Leverandørnr. TRCM1978531MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Matairesinol-d6
CAS 1346600-02-9
Molekylær formel C20H16D6O6
Mængde 1 mg
Synonym (3R,4R)-rel-Dihydro-3,4-bis[[4-hydroxy-3-methoxy(phenyl-d3)]methyl]-2(3H)-furanone, trans-Dihydro-3,4-bis[[4-hydroxy-3-methoxy(phenyl-d3)]methyl]-2(3H)-furanone
InChI formel InChI=1S/C20H22O6/c1-24-18-9-12(3-5-16(18)21)7-14-11-26-20(23)15(14)8-13-4-6-17(22)19(10-13)25-2/h3-6,9-10,14-15,21-22H,7-8,11H2,1-2H3/t14-,15+/m1/s1/i3D,4D,5D,6D,9D,10D
SMIL O=C1OC[C@@H](CC2=C([2H])C([2H])=C(O)C(OC)=C2[2H])[C@@H]1CC3=C([2H])C([2H])=C(O)C(OC)=C3[2H]
Molekylvægt (g/mol) 364.42