Få mere at vide
rac-Kavain, TRC
Artikelnummer. 30137678
Skift visning
Mængde:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137678 5 g
30008430 1 g
30004467 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137678 Leverandør Toronto Research Chemicals Leverandørnr. TRCK1454905G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3155-48-4
Molekylær formel C14 H14 O3
Molekylvægt (g/mol) 230.26
Synonym 2H-Pyran-2-one, 5,6-dihydro-4-methoxy-6-[(1E)-2-phenylethenyl]-, 2H-Pyran-2-one, 5,6-dihydro-4-methoxy-6-(2-phenylethenyl)-, (E)-(±)-, 2H-Pyran-2-one, 5,6-dihydro-4-methoxy-6-styryl-, (±)- (8CI), (±)-Kavain, (±)-Kawain, 2H-Pyran-2-one, 5,6-dihydro-4-methoxy-6-(2-phenylethenyl)-, (E)-, DL-Kawain, dl-Kavain
IUPAC navn 4-methoxy-2-[(E)-2-phenylethenyl]-2,3-dihydropyran-6-one
SMIL COC1=CC(=O)OC(C1)\C=C\c2ccccc2
Tekniske data
Kemisk navn eller materiale DL-Kavain
Mængde 5 g
InChI formel InChI=1S/C14H14O3/c1-16-13-9-12(17-14(15)10-13)8-7-11-5-3-2-4-6-11/h2-8,10,12H,9H2,1H3/b8-7+
Formel vægt 230.0943