Få mere at vide
rac Guaifenesin-d5, TRC
Artikelnummer. 30240723
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30240723 2.5 mg 2.5mg
30240686 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30240723 Leverandør Toronto Research Chemicals Leverandørnr. TRCG8105032.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1329563-41-8
Molekylær formel C10H9D5O4
Molekylvægt (g/mol) 203.25
Synonym 3-(o-Methoxyphenoxy)-1,2-propanediol-d5, Guaiacol Glyceryl-d5 Ether, Robitussin-d5
SMIL OC([2H])([2H])C(O)([2H])C([2H])([2H])OC1=C(OC)C=CC=C1
Tekniske data
Kemisk navn eller materiale rac Guaifenesin-d5
Renhedsgrad noter HPLC
Mængde 2.5 mg
InChI formel InChI=1S/C10H14O4/c1-13-9-4-2-3-5-10(9)14-7-8(12)6-11/h2-5,8,11-12H,6-7H2,1H3/i6D2,7D2,8D
Formel vægt 203.1206
Procent renhed >95%