missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
rac Fenfluramin-d5 Hydrochlorid, TRC
Specifications
Specifications
| Kemisk navn eller materiale | rac Fenfluramine-d5 Hydrochloride |
| CAS | 1216927-29-5 |
| Molekylær formel | C12 D5 H11 F3 N . Cl H |
| Mængde | 1 mg |
| Synonym | Fenfluramine Hydrochloride D5 (Ethyl D5), N-(Ethyl-d5)-α-methyl-3-(trifluoromethyl)benzeneethanamine hydrochloride, 3-(Trifluoromethyl)-N-(ethyl-d5)-α-methylphenethylamine hydrochloride, N-Pentadeuterioethyl-α-methyl-3-(trifluoromethyl)phenethylamine hydrochloride, N-(1,1,2,2,2-Pentadeuterioethyl)-3-[3-(trifluoromethyl)phenyl]propan-2-amine hydrochloride, Fenfluramine-d5 hydrochloride, Fenfluramine-D5 (N-ethyl-D5) hydrochloride |
| InChI formel | InChI=1S/C12H16F3N.ClH/c1-3-16-9(2)7-10-5-4-6-11(8-10)12(13,14)15;/h4-6,8-9,16H,3,7H2,1-2H3;1H/i1D3,3D2; |
| SMIL | Cl.[2H]C([2H])([2H])C([2H])([2H])NC(C)Cc1cccc(c1)C(F)(F)F |
| IUPAC navn | N-(1,1,2,2,2-pentadeuterioethyl)-1-[3-(trifluoromethyl)phenyl] propan-2-amin; hydrochlorid |
| Molekylvægt (g/mol) | 272.749 |
| Formel vægt | 272.132 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?