Få mere at vide
rac Felodipine-d3, TRC
Artikelnummer. 30405671
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30405671 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405671 Leverandør Toronto Research Chemicals Leverandørnr. TRCF2323771MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Felodipine O-methyl-d3
CAS 1189970-31-7
Molekylær formel C18 2H3 H16 Cl2 N O4
Mængde 1 mg
Synonym O5-Ethyl O3-(trideuteriomethyl) 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, Felodipine-d3, Felodipine-D3 (O-methyl-D3), 4-(2,3-Dichlorophenyl)-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic acid ethyl methyl-d3 ester
InChI formel InChI=1S/C18H19Cl2NO4/c1-5-25-18(23)14-10(3)21-9(2)13(17(22)24-4)15(14)11-7-6-8-12(19)16(11)20/h6-8,15,21H,5H2,1-4H3/i4D3
SMIL [2H]C([2H])([2H])OC(=O)C1=C(C)NC(=C(C1c2cccc(Cl)c2Cl)C(=O)OCC)C
IUPAC navn 3-O-ethyl 5-O-(trideuteriomethyl) 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Molekylvægt (g/mol) 387.27
Formel vægt 386.0879