Få mere at vide
rac erythro-Dihydro Bupropion-d9, TRC
Artikelnummer. 30371165
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30371165 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371165 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4486521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac erythro-Dihydro Bupropion-d9
CAS 1217684-77-9
Molekylær formel C13H11D9ClNO
Mængde 1 mg
Synonym (αS)-rel-3-Chloro-α-[(1R)-1-[(1,1-dimethylethyl)amino]ethyl]benzenemethanol-d9, rac erythro-Hydroxybupropion-d9
InChI formel InChI=1S/C13H20ClNO/c1-9(15-13(2,3)4)12(16)10-6-5-7-11(14)8-10/h5-9,12,15-16H,1-4H3/t9-,12-/m1/s1/i2D3,3D3,4D3
SMIL ClC1=CC([C@H](O)[C@@H](C)NC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=CC=C1
Molekylvægt (g/mol) 250.81
Formel vægt 250.1798