Få mere at vide
rac Dobutamine-d6 (Major) Hydrochloride, TRC
Artikelnummer. 30374010
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30374010 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30374010 Leverandør Toronto Research Chemicals Leverandørnr. TRCD49444825MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Dobutamine-d6 hydrochloride
CAS 1246818-96-1
Molekylær formel C18 2H6 H17 N O3 . Cl H
Mængde 25 mg
Synonym 4-[2-[[3-(4-Hydroxyphenyl)-1-(methyl-d3)propyl-1,2,2-d3]amino]ethyl]-1,2-benzenediol hydrochloride, Dobutamine-d6 hydrochloride, Dobutamine-D6 hydrochloride (2-aminobutyl-1,1,1,2,3,3-D6)
InChI formel InChI=1S/C18H23NO3.ClH/c1-13(2-3-14-4-7-16(20)8-5-14)19-11-10-15-6-9-17(21)18(22)12-15;/h4-9,12-13,19-22H,2-3,10-11H2,1H3;1H/i1D3,2D2,13D;
SMIL Cl.[2H]C([2H])([2H])C([2H])(NCCc1ccc(O)c(O)c1)C([2H])([2H])Cc2ccc(O)cc2
IUPAC navn 4-[2-[[1,1,1,2,3,3-hexadeuterio-4-(4-hydroxyphenyl)butan-2-yl]amino]ethyl]benzene-1,2-diol;hydrochloride
Molekylvægt (g/mol) 343.88
Formel vægt 343.1821