Få mere at vide
rac Desisopropyl Tolterodine-d7, TRC
Artikelnummer. 30363111
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30363111 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30363111 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29022710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Desisopropyl Tolterodine-d7
CAS 1346600-20-1
Molekylær formel C19H18D7NO
Mængde 10 mg
Synonym 4-Methyl-2-[3-[(1-methylethyl-d7)amino]-1-phenylpropyl]phenol, N-(Isopropyl-d7)-3-(2-hydroxy-5-methylphenyl)-3-phenylpropylamine
InChI formel InChI=1S/C19H25NO/c1-14(2)20-12-11-17(16-7-5-4-6-8-16)18-13-15(3)9-10-19(18)21/h4-10,13-14,17,20-21H,11-12H2,1-3H3/i1D3,2D3,14D
SMIL [2H]C([2H])([2H])C([2H])(NCCC(c1ccccc1)c2cc(C)ccc2O)C([2H])([2H])[2H]
IUPAC navn 2-[3-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)-1-phenylpropyl]-4-methylphenol
Molekylvægt (g/mol) 290.45
Formel vægt 290.2376