missing translation for 'onlineSavingsMsg'
Få mere at vide

rac Cycloserine-15N,d3, TRC

Artikelnummer. 30355615
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30355615 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30355615 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9888021MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale rac Cycloserine-15N,d3
CAS 1219176-26-7
Renhedsgrad noter HPLC
Molekylær formel C3H3D3N15NO2
Mængde 1 mg
Synonym 4-Amino-3-isoxazolidinone-15N,d3, (+/-)-3-Oxoisooxazolidin-4-amine-15N,d3, DL-Cycloserine-15N,d3
InChI formel InChI=1S/C3H6N2O2/c4-2-1-7-5-3(2)6/h2H,1,4H2,(H,5,6)/i1D2,2D,5+1
SMIL [2H]C1([2H])C(N)([2H])C([15NH]O1)=O
IUPAC navn 4-amino-4,5,5-trideuterio-(215N)1,2-oxazolidin-3-one
Molekylvægt (g/mol) 106.1
Procent renhed >95
Vis mere Vis mindre
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.