Få mere at vide
rac-Cotinine, TRC
Artikelnummer. 30179926
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179926 10 mg
30179717 25 mg
30180333 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179926 Leverandør Toronto Research Chemicals Leverandørnr. TRCC72500410MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 15569-85-4
Molekylær formel C10 H12 N2 O
Molekylvægt (g/mol) 176.22
Synonym rac-Cotinine
IUPAC navn 1-methyl-5-pyridin-3-ylpyrrolidin-2-one
SMIL CN1C(CCC1=O)c2cccnc2
Tekniske data
Kemisk navn eller materiale rac Cotinine
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C10H12N2O/c1-12-9(4-5-10(12)13)8-3-2-6-11-7-8/h2-3,6-7,9H,4-5H2,1H3
Formel vægt 176.095
Procent renhed >95%