missing translation for 'onlineSavingsMsg'
Få mere at vide

rac Clopidogrel-d4 Hydrogen Sulfate, TRC

Artikelnummer. 30349053
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30349053 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30349053 Leverandør Toronto Research Chemicals Leverandørnr. TRCC58725210MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale (RS)-Clopidogrel 2-chlorophenyl-d4 hydrogen sulfate
CAS 1219274-96-0
Renhedsgrad noter HPLC
Molekylær formel C16 2H4 H12 Cl N O2 S . H2 O4 S
Mængde 10 mg
Synonym Methyl 2-(2-chloro-3,4,5,6-tetradeuteriophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate sulfuric acid, rac-Clopidogrel-d4 hydrogen sulfate, (RS)-Methyl (2-chlorophenyl-3,4,5,6-d4)(6,7-dihydro-4H-thieno[3,2-c]pyrid-5-yl)acetate hydrogen sulfate salt, Methyl 2-(2-Chlorophenyl-3,4,5,6-d4)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate hydrogen sulfate, (RS)-Clopidogrel-d4 hydrogen sulfate, (RS)-Clopidogrel-D4 hydrogen sulfate (2-chlorophenyl-D4)
InChI formel InChI=1S/C16H16ClNO2S.H2O4S/c1-20-16(19)15(12-4-2-3-5-13(12)17)18-8-6-14-11(10-18)7-9-21-14;1-5(2,3)4/h2-5,7,9,15H,6,8,10H2,1H3;(H2,1,2,3,4)/i2D,3D,4D,5D;
SMIL [2H]c1c([2H])c([2H])c(C(N2CCc3sccc3C2)C(=O)OC)c(Cl)c1[2H].OS(=O)(=O)O
IUPAC navn methyl 2-(2-chloro-3,4,5,6-tetradeuteriophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate;sulfuric acid
Molekylvægt (g/mol) 423.92
Formel vægt 423.0515
Procent renhed >95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.