Få mere at vide
rac-Clopidogrel-d4 Carboxylic Acid, TRC
Artikelnummer. 30354334
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30354334 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30354334 Leverandør Toronto Research Chemicals Leverandørnr. TRCC58724210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (+/-)- Clopidogrel-d4 Carboxylic Acid
CAS 1246814-52-7
Renhedsgrad noter HPLC
Molekylær formel C15 2H4 H10 Cl N O2 S
Mængde 10 mg
InChI formel InChI=1S/C15H14ClNO2S/c16-12-4-2-1-3-11(12)14(15(18)19)17-7-5-13-10(9-17)6-8-20-13/h1-4,6,8,14H,5,7,9H2,(H,18,19)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c([2H])c(C(N2CCc3sccc3C2)C(=O)O)c(Cl)c1[2H]
IUPAC navn 2-(2-chloro-3,4,5,6-tetradeuteriophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetic acid
Molekylvægt (g/mol) 311.82
Formel vægt 311.0685
Procent renhed >95