Få mere at vide
rac Clopidogrel-13C,d3 Hydrogen Sulfate, TRC
Artikelnummer. 30362120
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30362120 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30362120 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5872532.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Clopidogrel-13C,d3 Hydrogen Sulfate
CAS 1246814-55-0
Molekylær formel 13C C15 2H3 H13 Cl N O2 S . H2 O4 S
Mængde 2.5 mg
Synonym Trideuteriomethyl-13C 2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate sulfuric acid, rac-Clopidogrel-13C,d3 hydrogen sulfate, (RS)-Methyl-13C,d3 (2-chlorophenyl)(6,7-dihydro-4H-thieno[3,2-c]pyrid-5-yl)acetate hydrogen sulfate salt, Methyl-13C,d3 2-(2-Chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate hydrogen sulfate, (RS)-Clopidogrel-13C,d3 hydrogen sulfate, (RS)-Clopidogrel-13C,D3 hydrogen sulfate (O-methyl-13C,D3)
InChI formel InChI=1S/C16H16ClNO2S.H2O4S/c1-20-16(19)15(12-4-2-3-5-13(12)17)18-8-6-14-11(10-18)7-9-21-14;1-5(2,3)4/h2-5,7,9,15H,6,8,10H2,1H3;(H2,1,2,3,4)/i1+1D3;
SMIL [2H][13C]([2H])([2H])OC(=O)C(N1CCc2sccc2C1)c3ccccc3Cl.OS(=O)(=O)O
IUPAC navn sulfuric acid;trideuterio(1^{13}C)methyl 2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate
Molekylvægt (g/mol) 423.91
Formel vægt 423.0486