Få mere at vide
rac Cinacalcet-d3 Hydrochloride, TRC
Artikelnummer. 30364202
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30364202 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30364202 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4418021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Cinacalcet-d3 Hydrochloride
CAS 1185097-33-9
Molekylær formel C22 2H3 H19 F3 N . Cl H
Mængde 1 mg
Synonym N-(2,2,2-Trideuterio-1-naphthalen-1-ylethyl)-3-[3-(trifluoromethyl)phenyl]propan-1-amine hydrochloride, rac Cinacalcet-d3 hydrochloride, α-Trideuteriomethyl-N-[3-[3-(trifluoromethyl)phenyl]propyl]-1-naphthalenemethanamine hydrochloride, (RS)-Cinacalcet D3 hydrochloride (methyl D3), N-[2,2,2-Trideuterio-1-(1-naphthyl)ethyl]-3-[3-(trifluoromethyl)phenyl]propan-1-amine hydrochloride
InChI formel InChI=1S/C22H22F3N.ClH/c1-16(20-13-5-10-18-9-2-3-12-21(18)20)26-14-6-8-17-7-4-11-19(15-17)22(23,24)25;/h2-5,7,9-13,15-16,26H,6,8,14H2,1H3;1H/i1D3;
SMIL Cl.[2H]C([2H])([2H])C(NCCCc1cccc(c1)C(F)(F)F)c2cccc3ccccc23
IUPAC navn N-(2,2,2-trideuterio-1-naphthalen-1-ylethyl)-3-[3-(trifluoromethyl)phenyl]propan-1-amine;hydrochloride
Molekylvægt (g/mol) 396.89
Formel vægt 396.1659