Få mere at vide
rac Carprofen-d3, TRC
Artikelnummer. 30363504
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30363504 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30363504 Leverandør Toronto Research Chemicals Leverandørnr. TRCC18435210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Carprofen-d3
CAS 1173019-42-5
Molekylær formel C15 2H3 H9 Cl N O2
Mængde 10 mg
Synonym 2-(6-Chloro-9H-carbazol-2-yl)-3,3,3-trideuteriopropanoic acid, Carprofen-d3, Carprofen D3 (methyl D3), rac Carprofen-d3
InChI formel InChI=1S/C15H12ClNO2/c1-8(15(18)19)9-2-4-11-12-7-10(16)3-5-13(12)17-14(11)6-9/h2-8,17H,1H3,(H,18,19)/i1D3
SMIL [2H]C([2H])([2H])C(C(=O)O)c1ccc2c(c1)[nH]c3ccc(Cl)cc23
IUPAC navn 2-(6-chloro-9H-carbazol-2-yl)-3,3,3-trideuteriopropanoic acid
Molekylvægt (g/mol) 276.73
Formel vægt 276.0745