Få mere at vide
rac Bopindolol, TRC
Artikelnummer. 30138558
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138558 50 mg
30010977 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138558 Leverandør Toronto Research Chemicals Leverandørnr. TRCB67540050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 62658-63-3
Molekylær formel C23 H28 N2 O3
Molekylvægt (g/mol) 380.48
Synonym 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-[(2-methyl-1H-indol-4-yl)oxy]-, 2-benzoate, 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-[(2-methyl-1H-indol-4-yl)oxy]-, benzoate (ester) (9CI), 2-Propanol, 1-[(1,1-dimethylethyl)amino]-3-[(2-methyl-1H-indol-4-yl)oxy]-, benzoate (ester), (±)-, (±)-Bopindolol, Bopindolol, rac Bopindolol
IUPAC navn [1-(tert-butylamino)-3-[(2-methyl-1H-indol-4-yl)oxy]propan-2-yl] benzoate
SMIL Cc1cc2c(OCC(CNC(C)(C)C)OC(=O)c3ccccc3)cccc2[nH]1
Tekniske data
Kemisk navn eller materiale rac Bopindolol
Mængde 50 mg
InChI formel InChI=1S/C23H28N2O3/c1-16-13-19-20(25-16)11-8-12-21(19)27-15-18(14-24-23(2,3)4)28-22(26)17-9-6-5-7-10-17/h5-13,18,24-25H,14-15H2,1-4H3
Formel vægt 380.21