missing translation for 'onlineSavingsMsg'
Få mere at vide

rac Bepotastine Ethyl Ester, TRC

Artikelnummer. 30356654
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30356654 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30356654 Leverandør Toronto Research Chemicals Leverandørnr. TRCC387455250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale rac Bepotastine Ethyl Ester
CAS 125602-69-9
Molekylær formel C23H29ClN2O3
Mængde 250 mg
InChI formel InChI=1S/C23H29ClN2O3/c1-2-28-22(27)7-5-15-26-16-12-20(13-17-26)29-23(21-6-3-4-14-25-21)18-8-10-19(24)11-9-18/h3-4,6,8-11,14,20,23H,2,5,7,12-13,15-17H2,1H3
SMIL CCOC(=O)CCCN1CCC(CC1)OC(c2ccc(Cl)cc2)c3ccccn3
IUPAC navn ethyl 4-[4-[(4-chlorophenyl)-pyridin-2-ylmethoxy]piperidin-1-yl]butanoate
Molekylvægt (g/mol) 416.94
Formel vægt 416.1867
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.