Få mere at vide
rac Bendroflumethiazide-d5, TRC
Artikelnummer. 30342589
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30342589 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30342589 Leverandør Toronto Research Chemicals Leverandørnr. TRCB1335025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Bendroflumethiazide-d5
CAS 1330183-13-5
Renhedsgrad noter HPLC
Molekylær formel C15H9D5F3N3O4S2
Mængde 5 mg
Synonym 3,4-Dihydro-3-(phenylmethyl-d5)-6-(trifluoromethyl)-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-Dioxide, 3-(Benzyl-d5)-3,4-dihydro-6-(trifluoromethyl)-2H-1,2,4- benzothiadiazine-7-sulfonamide 1,1-Dioxide, Aprinox-d5, Be 724A-d5, Bendrofluazide-d5, (+/-)-Bendroflumethiazide-d5, Bristuron-d5, Centyl-d5, Corzide-d5, FT 81-d5, Flumersil-d5, Naturine-d5, Sinesaline-d5, Sodiuretic-d5, Thiazidico-d5, Urlea-d5
InChI formel InChI=1S/C15H14F3N3O4S2/c16-15(17,18)10-7-11-13(8-12(10)26(19,22)23)27(24,25)21-14(20-11)6-9-4-2-1-3-5-9/h1-5,7-8,14,20-21H,6H2,(H2,19,22,23)/i1D,2D,3D,4D,5D
SMIL O=S1(C2=CC(S(N)(=O)=O)=C(C(F)(F)F)C=C2NC(CC3=C([2H])C([2H])=C([2H])C([2H])=C3[2H])N1)=O
IUPAC navn 1,1-dioxo-3-[(2,3,4,5,6-pentadeuteriophenyl)methyl]-6-(trifluoromethyl)-3,4-dihydro-2H-1λ6,2,4-benzothiadiazine-7-sulfonamide
Molekylvægt (g/mol) 426.45
Procent renhed >95