Få mere at vide
rac Atomoxetine-d5 Hydrochloride, TRC
Artikelnummer. 30337662
Skift visning
Mængde:
0.5 mg
Artikelnummer. Mængde
30337662 0.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30337662 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7913920.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Atomoxetine-d5 Hydrochloride
CAS 1398065-95-6
Molekylær formel C17 D5 H16 N O . Cl H
Mængde 0.5 mg
Synonym N-Methyl-3-(pentadeuteriophenyl)-3-(2-methylphenoxy)propan-1-amine hydrochloride (1:1), 3-(2-Methylphenoxy)-3-(phenyl-d5)-N-methylpropan-1-amine hydrochloride, N-Methyl-3-(2-methylphenoxy)-3-(phenyl-d5)propylamine monohydrochloride, rac Atomoxetine-d5 hydrochloride, (±)-Atomoxetine-d5 hydrochloride, (RS)-Atomoxetine-D5 (phenyl-D5) hydrochloride
InChI formel InChI=1S/C17H21NO.ClH/c1-14-8-6-7-11-16(14)19-17(12-13-18-2)15-9-4-3-5-10-15;/h3-11,17-18H,12-13H2,1-2H3;1H/i3D,4D,5D,9D,10D;
SMIL Cl.[2H]c1c([2H])c([2H])c(C(CCNC)Oc2ccccc2C)c([2H])c1[2H]
IUPAC navn N-methyl-3-(2-methylphenoxy)-3-(2,3,4,5,6-pentadeuteriophenyl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 296.846
Formel vægt 296.17