Få mere at vide
rac Ambrisentan-d3 Methyl Ester, TRC
Artikelnummer. 30315942
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30315942 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315942 Leverandør Toronto Research Chemicals Leverandørnr. TRCA5758672.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac Ambrisentan-d3 Methyl Ester
CAS 1329837-75-3
Molekylær formel C23H21D3N2O4
Mængde 2.5 mg
Synonym α-[(4,6-Dimethyl-2-pyrimidinyl)oxy]-β-methoxy-β-phenyl-benzenepropanoic Acid-d3 Methyl Ester
InChI formel InChI=1S/C23H24N2O4/c1-16-15-17(2)25-22(24-16)29-20(21(26)27-3)23(28-4,18-11-7-5-8-12-18)19-13-9-6-10-14-19/h5-15,20H,1-4H3/i4D3
SMIL CC1=CC(C)=NC(OC(C(OC)=O)C(C2=CC=CC=C2)(OC([2H])([2H])[2H])C3=CC=CC=C3)=N1
IUPAC navn methyl 2-(4,6-dimethylpyrimidin-2-yl)oxy-3,3-diphenyl-3-(trideuteriomethoxy)propanoate
Molekylvægt (g/mol) 395.47