Få mere at vide
rac Alpha-Lipoic Acid-d5, TRC
Artikelnummer. 30480091
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30480091 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30480091 Leverandør Toronto Research Chemicals Leverandørnr. TRCL4687521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac a-Lipoic Acid-d5
CAS 1189471-66-6
Molekylær formel C8H9D5O2S2
Mængde 1 mg
Synonym Alpha Lipoic Acid D5
InChI formel InChI=1S/C8H14O2S2/c9-8(10)4-2-1-3-7-5-6-11-12-7/h7H,1-6H2,(H,9,10)/i3D2,5D2,7D
SMIL [2H]C([2H])(CCCC(=O)O)C1([2H])SSCC1([2H])[2H]
Molekylvægt (g/mol) 211.36