Få mere at vide
rac 8-Hydroxy Efavirenz-d4, TRC
Artikelnummer. 30405159
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30405159 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405159 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94182710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac 8-Hydroxy Efavirenz-d4
CAS 1246812-30-5
Renhedsgrad noter HPLC
Molekylær formel C14H5D4ClF3NO3
Mængde 10 mg
Synonym 6-Chloro-4-(cyclopropyl-d4-ethynyl)-1,4-dihydro-8-hydroxy-4-(trifluoromethyl)-2H-3,1-benzoxazin-2-one, 8-Hydroxyefavirenz
InChI formel InChI=1S/C14H9ClF3NO3/c15-8-5-9-11(10(20)6-8)19-12(21)22-13(9,14(16,17)18)4-3-7-1-2-7/h5-7,20H,1-2H2,(H,19,21)/i1D2,2D2
SMIL ClC1=CC(C(C#CC2C([2H])([2H])C2([2H])[2H])(C(F)(F)F)OC(N3)=O)=C3C(O)=C1
Molekylvægt (g/mol) 335.7
Formel vægt 335.0474
Procent renhed >95