missing translation for 'onlineSavingsMsg'
Få mere at vide

rac 7,14-Dihydroxy Efavirenz, TRC

Artikelnummer. 30370589
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30370589 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30370589 Leverandør Toronto Research Chemicals Leverandørnr. TRCD45297010MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale rac 7,14-Dihydroxy Efavirenz
CAS 1702668-54-9
Molekylær formel C14H9ClF3NO4
Mængde 10 mg
Synonym 6-Chloro-1,4-dihydro-7-hydroxy-4-[2-(1-hydroxycyclopropyl)ethynyl]-4-(trifluoromethyl)-2H-3,1-benzoxazin-2-one
InChI formel InChI=1S/C14H9ClF3NO4/c15-8-5-7-9(6-10(8)20)19-11(21)23-13(7,14(16,17)18)4-3-12(22)1-2-12/h5-6,20,22H,1-2H2,(H,19,21)/t13-/m0/s1
SMIL ClC1=CC(C(C#CC2(O)CC2)(C(F)(F)F)OC(N3)=O)=C3C=C1O
IUPAC navn 6-chloro-7-hydroxy-4-((1-hydroxycyclopropyl)ethynyl)-4-(trifluoromethyl)-1,4-dihydro-2H-benzo[d][1,3]oxazin-2-one
Molekylvægt (g/mol) 347.67
Formel vægt 347.02
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.