missing translation for 'onlineSavingsMsg'
Få mere at vide

rac 6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol, TRC

Artikelnummer. 30405339
Change view
Click to view available options
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30405339 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405339 Supplier Toronto Research Chemicals Supplier No. TRCF590980250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale rac 6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol
CAS 99199-62-9
Molekylær formel C10H11FO2
Mængde 250 mg
InChI formel InChI=1S/C10H11FO2/c11-8-2-4-10-7(5-8)1-3-9(6-12)13-10/h2,4-5,9,12H,1,3,6H2
SMIL OCC1CCc2cc(F)ccc2O1
IUPAC navn (6-fluoro-3,4-dihydro-2H-chromen-2-yl)methanol
Molekylvægt (g/mol) 182.19
Formel vægt 182.0743
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.