Få mere at vide
rac 5-Carboxy Desisopropyl Tolterodine-d7, TRC
Artikelnummer. 30367913
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30367913 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30367913 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1778871MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac 5-Carboxy Desisopropyl Tolterodine-d6
CAS 1189868-60-7
Molekylær formel C19H16D7NO3
Mængde 1 mg
InChI formel InChI=1S/C19H23NO3/c1-13(2)20-11-10-16(14-6-4-3-5-7-14)17-12-15(19(22)23)8-9-18(17)21/h3-9,12-13,16,20-21H,10-11H2,1-2H3,(H,22,23)/i1D3,2D3,13D
SMIL [2H]C([2H])([2H])C([2H])(NCCC(c1ccccc1)c2cc(ccc2O)C(=O)O)C([2H])([2H])[2H]
IUPAC navn 3-[3-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)-1-phenylpropyl]-4-hydroxybenzoic acid
Molekylvægt (g/mol) 320.43
Formel vægt 320.2117