Få mere at vide
rac 5-Carboxy Desisopropyl Tolterodine-d7 Methyl Ester, TRC
Artikelnummer. 30361272
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30361272 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361272 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1779372.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac 5-Carboxy Desisopropyl Tolterodine-d7 Methyl Ester
CAS 1794780-63-4
Molekylær formel C20H18D7NO3
Mængde 2.5 mg
Synonym 4-Hydroxy-3-[3-[(1-methylethyl-d7)amino]-1-phenylpropyl]-benzoic Acid Methyl Ester
InChI formel InChI=1S/C20H25NO3/c1-14(2)21-12-11-17(15-7-5-4-6-8-15)18-13-16(20(23)24-3)9-10-19(18)22/h4-10,13-14,17,21-22H,11-12H2,1-3H3/i1D3,2D3,14D
SMIL [2H]C([2H])([2H])C([2H])(NCCC(c1ccccc1)c2cc(ccc2O)C(=O)OC)C([2H])([2H])[2H]
IUPAC navn methyl 3-[3-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)-1-phenylpropyl]-4-hydroxybenzoate
Molekylvægt (g/mol) 334.46
Formel vægt 334.2274