Få mere at vide
rac 5,6-Dehydro Pregabalin, TRC
Artikelnummer. 17922495
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17922495 10 mg
17930355 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17922495 Leverandør Toronto Research Chemicals Leverandørnr. TRCD23005510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1136478-30-2
Molekylær formel C8H15NO2
Molekylvægt (g/mol) 157.21
Synonym 5-Hexenoic acid, 3-(aminomethyl)-5-methyl-, 3-(Aminomethyl)-5-methyl-5-hexenoic acid, 3-(Aminomethyl)-5-methylhex-5-enoic acid, (RS)-5,6-Dehydropregabalin, rac 5,6-Dehydro Pregabalin
SMIL CC(=C)CC(CN)CC(=O)O
Tekniske data
Kemisk navn eller materiale rac 5,6-Dehydro Pregabalin
InChI formel InChI=1S/C8H15NO2/c1-6(2)3-7(5-9)4-8(10)11/h7H,1,3-5,9H2,2H3,(H,10,11)
Mængde 10 mg
Formel vægt 157.1103