Få mere at vide
rac 4-Hydroxy-3-methoxyphenylethylene Glycol-d3, TRC
Artikelnummer. 30408125
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30408125 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30408125 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94634225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac 4-Hydroxy-3-methoxyphenylethylene Glycol-d3
CAS 74495-72-0
Molekylær formel C9 2H3 H9 O4
Mængde 25 mg
Synonym 1-(4-Hydroxy-3-(methoxy-d3)phenyl)-1,2-ethanediol, [3-(Methoxy-d3)-4-hydroxyphenyl]ethylene Glycol, [4-Hydroxy-3-(methoxy-d3)phenyl]ethylene Glycol, 4-Hydroxy-3-(methoxy-d3)phenyl Glycol, HMPG-d3, MHPG-d3, MOPEG-d3, Vanilethanediol-d3, Vanylglycol-d3
InChI formel InChI=1S/C9H12O4/c1-13-9-4-6(8(12)5-10)2-3-7(9)11/h2-4,8,10-12H,5H2,1H3/i1D3
SMIL [2H]C([2H])([2H])Oc1cc(ccc1O)C(O)CO
IUPAC navn 1-[4-hydroxy-3-(trideuteriomethoxy)phenyl]ethane-1,2-diol
Molekylvægt (g/mol) 187.21
Formel vægt 187.0924