Få mere at vide
rac 4,5-Dehydro Pregabalin, TRC
Artikelnummer. 30003790
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003790 5 mg
30011092 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30003790 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2300505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 216576-74-8
Molekylær formel C8 H15 N O2
Molekylvægt (g/mol) 157.21
Synonym 4-Hexenoic acid, 3-(aminomethyl)-5-methyl-, 3-(Aminomethyl)-5-methyl-4-hexenoic acid
IUPAC navn 3-(aminomethyl)-5-methylhex-4-enoic acid
SMIL CC(=CC(CN)CC(=O)O)C
Tekniske data
Kemisk navn eller materiale rac 4,5-Dehydro Pregabalin
Renhedsgrad noter HPLC
InChI formel InChI=1S/C8H15NO2/c1-6(2)3-7(5-9)4-8(10)11/h3,7H,4-5,9H2,1-2H3,(H,10,11)
Mængde 5 mg
Formel vægt 157.1103
Procent renhed >95%