Få mere at vide
rac 4-(3-Aminobutyl)phenol-d6, TRC
Artikelnummer. 30310267
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30310267 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310267 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60250225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac 4-(3-Aminobutyl)phenol-d6
CAS 1189890-45-6
Molekylær formel C10H9D6NO
Mængde 25 mg
Synonym 4-(3-Amino-2,2,3,4,4,4-hexadeuteriobutyl)phenol, 4-(3-Aminobutyl)phenol-D6 (butyl-2,2,3,4,4,4-D6)
InChI formel InChI=1S/C10H15NO/c1-8(11)2-3-9-4-6-10(12)7-5-9/h4-8,12H,2-3,11H2,1H3/i1D3,2D2,8D
SMIL [2H]C([2H])([2H])C([2H])(N)C([2H])([2H])Cc1ccc(O)cc1
IUPAC navn 4-(3-amino-2,2,3,4,4,4-hexadeuteriobutyl)phenol
Molekylvægt (g/mol) 171.27
Formel vægt 171.153