missing translation for 'onlineSavingsMsg'
Learn More

rac-3-Octadecanamido-2-ethoxypropan-1-ol Phosphocholine, TRC

Product Code. 30423941
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30423941 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423941 Supplier Toronto Research Chemicals Supplier No. TRCO23400010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale rac-3-Octadecanamido-2-Ethoxypropan-1-ol Phosphocholine
CAS 112989-02-3
Molekylær formel C28 H59 N2 O6 P
Mængde 10 mg
Synonym 3,5-Dioxa-9-aza-4-phosphaheptacosan-1-aminium','7-ethoxy-4-hydroxy-N,N,N-trimethyl-10-oxo-','inner salt','4-oxide, 3,5-Dioxa-9-aza-4-phosphaheptacosan-1-aminium','7-ethoxy-4-hydroxy-N,N,N-trimethyl-10-oxo-','inner salt','4-oxide','(±)-, CP 51, rac-3-Octadecanamido-2-ethoxypropan-1-ol Phosphocholine
InChI formel InChI=1S/C28H59N2O6P/c1-6-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-28(31)29-25-27(34-7-2)26-36-37(32,33)35-24-23-30(3,4)5/h27H,6-26H2,1-5H3,(H-,29,31,32,33)
SMIL CCCCCCCCCCCCCCCCCC(=O)NCC(COP(=O)([O-])OCC[N+](C)(C)C)OCC
IUPAC navn [2-ethoxy-3-(octadecanoylamino)propyl] 2-(trimethylazaniumyl)ethyl phosphate
Molekylvægt (g/mol) 550.75
Formel vægt 550.4111
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.