Få mere at vide
rac 3-O-Methyl DOPA, TRC
Artikelnummer. 30004209
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004209 25 mg
17942265 100 mg
17971905 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30004209 Leverandør Toronto Research Chemicals Leverandørnr. TRCM30380525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7636-26-2
Molekylær formel C10 H13 N O4
Molekylvægt (g/mol) 211.21
Synonym (2RS)-2-Amino-3-(4-hydroxy-3-methoxyphenyl)propanoic Acid, Levodopa Imp. C (EP), 3-Methoxy-DL-tyrosine, (DL)-3-O-Methyldopa, Levodopa Related Compound B, 3-methoxytyrosine
IUPAC navn 2-amino-3-(4-hydroxy-3-methoxyphenyl)propanoic acid
SMIL COc1cc(CC(N)C(=O)O)ccc1O
Tekniske data
Kemisk navn eller materiale 3-Methoxy-DL-tyrosine
Renhedsgrad noter HPLC
InChI formel InChI=1S/C10H13NO4/c1-15-9-5-6(2-3-8(9)12)4-7(11)10(13)14/h2-3,5,7,12H,4,11H2,1H3,(H,13,14)
Mængde 25 mg
Formel vægt 211.0845
Procent renhed >95%