Få mere at vide
rac-3-Aminoisobutyric Acid-d3 Hydrochloride, TRC
Artikelnummer. 30312339
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30312339 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312339 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6115875MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac-3-Aminoisobutyric Acid-d3 Hydrochloride
CAS 1346602-62-7
Molekylær formel C4H7D3ClNO2
Mængde 5 mg
Synonym 3-Amino-2-(methyl-d3)propanoic Acid Hydrochloride, 3-Amino-2-(methyl-d3)propanoic Acid-d3 Hydrochloride, 2-(Methyl-d3)-β-alanine-d3 Hydrochloride, (±)-3-Aminoisobutyric-d3 Acid Hydrochloride, DL-β-Aminoisobutyric-d3 Acid Hydrochloride
InChI formel InChI=1S/C4H9NO2.ClH/c1-3(2-5)4(6)7;/h3H,2,5H2,1H3,(H,6,7);1H/i1D3;
SMIL NCC(C([2H])([2H])[2H])C(O)=O.Cl
IUPAC navn 2-(aminomethyl)-3,3,3-trideuteriopropanoic acid;hydrochloride
Molekylvægt (g/mol) 142.6