Få mere at vide
rac 2-Oleoyl-3-chloropropanediol-d5, 95%, TRC
Artikelnummer. 30425568
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30425568 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30425568 Leverandør Toronto Research Chemicals Leverandørnr. TRCO52681225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac 2-Oleoyl-3-chloropropanediol-d5
CAS 1331669-45-4
Molekylær formel C21H34D5ClO3
Mængde 25 mg
Synonym (9Z)-9-Octadecenoic Acid 2-Chloro-1-(hydroxymethyl)ethyl Ester-d5, (1-Chloro-1,1,2,3,3-pentadeuterio-3-hydroxypropan-2-yl) Octadec-9-enoate
InChI formel InChI=1S/C21H39ClO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-20(18-22)19-23/h9-10,20,23H,2-8,11-19H2,1H3/b10-9-/i18D2,19D2,20D
SMIL [2H]C([2H])(O)C([2H])(OC(=O)CCCCCCC\C=C/CCCCCCCC)C([2H])([2H])Cl
IUPAC navn (1-chloro-1,1,2,3,3-pentadeuterio-3-hydroxypropan-2-yl) (Z)-octadec-9-enoate
Molekylvægt (g/mol) 380.02
Formel vægt 379.2902