Få mere at vide
rac-2-Linoleoyl-3-chloropropanediol-d5, 95%, TRC
Artikelnummer. 30475731
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30475731 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30475731 Leverandør Toronto Research Chemicals Leverandørnr. TRCL4678072.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac-2-Linoleoyl-3-chloropropanediol-d5
CAS 1331636-39-5
Molekylær formel C21H32D5ClO3
Mængde 2.5 mg
Synonym 3-Chloro-1,2-propanediol-d5 2-Linoleate
InChI formel InChI=1S/C21H37ClO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-20(18-22)19-23/h6-7,9-10,20,23H,2-5,8,11-19H2,1H3/b7-6-,10-9-/i18D2,19D2,20D
SMIL [2H]C([2H])(O)C([2H])(OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)C([2H])([2H])Cl
IUPAC navn (1-chloro-1,1,2,3,3-pentadeuterio-3-hydroxypropan-2-yl) (9Z,12Z)-octadeca-9,12-dienoate
Molekylvægt (g/mol) 378
Formel vægt 377.2745