Få mere at vide
rac 2-Ethenyl-4,6-dimethoxy-benzoic Acid 1-Methyl-5-oxo-9-decen-1-yl Ester-d6, TRC
Artikelnummer. 30379197
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30379197 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30379197 Leverandør Toronto Research Chemicals Leverandørnr. TRCE89040725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac 2-Ethenyl-4,6-dimethoxy-benzoic Acid 1-Methyl-5-oxo-9-decen-1-yl Ester-d6
CAS 1246819-89-5
Molekylær formel C24H24D6O5
Mængde 25 mg
Synonym 2-Ethenyl-4,6-dimethoxy-benzoic Acid 1-Methyl-5-oxo-9-decen-1-yl Ester-d6
InChI formel InChI=1S/C22H30O5/c1-6-8-9-12-18(23)13-10-11-16(3)27-22(24)21-17(7-2)14-19(25-4)15-20(21)26-5/h6-7,14-16H,1-2,8-13H2,3-5H3/i3D3,11D2,16D
SMIL [2H]C([2H])([2H])C([2H])(OC(=O)c1c(OC)cc(OC)cc1C=C)C([2H])([2H])CCC(=O)CCCC=C
IUPAC navn (1,1,1,2,3,3-hexadeuterio-6-oxoundec-10-en-2-yl) 2-ethenyl-4,6-dimethoxybenzoate
Molekylvægt (g/mol) 380.51
Formel vægt 380.247