Få mere at vide
rac 1-Palmitoyl-2-oleoyl-3-chloropropanediol, TRC
Artikelnummer. 30012311
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012311 10 mg
30008815 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30012311 Leverandør Toronto Research Chemicals Leverandørnr. TRCP16050010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1363153-60-9
Molekylær formel C37 H69 Cl O4
Molekylvægt (g/mol) 613.39
IUPAC navn (Z)-18-(chloromethyl)-17,18-dihydroxyhexatriacont-27-ene-16,19-dione
SMIL CCCCCCCCCCCCCCCC(=O)C(O)C(O)(CCl)C(=O)CCCCCCC\C=C/CCCCCCCC
Tekniske data
Kemisk navn eller materiale 1-Palmitoyl-2-oleoyl-3-chloropropanediol
Mængde 10 mg
InChI formel InChI=1S/C37H69ClO4/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-35(40)37(42,33-38)36(41)34(39)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h17-18,36,41-42H,3-16,19-33H2,1-2H3/b18-17-
Formel vægt 612.4884