Få mere at vide
rac-1-Palmitoyl-2-linoleoyl-3-chloropropanediol-d5, TRC
Artikelnummer. 30409930
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30409930 2.5 mg 2.5mg
30425147 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30409930 Leverandør Toronto Research Chemicals Leverandørnr. TRCP1589522.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1246833-66-8
Molekylær formel C37H62D5ClO4
Molekylvægt (g/mol) 616.41
SMIL O=C(CCCCCCCCCCCCCCC)OC([2H])([2H])C(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)([2H])C([2H])([2H])Cl
Tekniske data
Kemisk navn eller materiale rac-1-Palmitoyl-2-linoleoyl-3-chloropropanediol-d5
Mængde 2.5 mg
InChI formel InChI=1S/C37H67ClO4/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-37(40)42-35(33-38)34-41-36(39)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h11,13,17-18,35H,3-10,12,14-16,19-34H2,1-2H3/b13-11+,18-17+
Formel vægt 615.5042