Få mere at vide
rac-1-Palmitoyl-2-linolenoyl-3-chloropropanediol-d5, TRC
Artikelnummer. 30422394
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30422394 25 mg
30427059 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30422394 Leverandør Toronto Research Chemicals Leverandørnr. TRCP15885225MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1246834-24-1
Molekylær formel C37H60D5ClO4
Molekylvægt (g/mol) 614.39
Synonym 9,12,15-Octadecatrienoic Acid 1-(Chloromethyl)-2-[(1-oxohexadecyl)oxy]ethyl Ester-d5
SMIL O=C(CCCCCCCCCCCCCCC)OC([2H])([2H])C(OC(CCCCCCC/C=C\C/C=C\C/C=C/CC)=O)([2H])C([2H])([2H])Cl
Tekniske data
Kemisk navn eller materiale rac-1-Palmitoyl-2-linolenoyl-3-chloropropanediol-d5
Mængde 25 mg
Formel vægt 613.4885