Få mere at vide
rac 1-Oleoyl-3-chloropropanediol-d5, TRC
Artikelnummer. 30426499
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30426499 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30426499 Leverandør Toronto Research Chemicals Leverandørnr. TRCO52680225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale rac 1-Oleoyl-3-chloropropanediol-d5
CAS 1246834-03-6
Molekylær formel C21H34D5ClO3
Mængde 25 mg
Synonym (9Z)-9-Octadecenoic Acid 3-Chloro-2-hydroxypropyl-d5 Ester, Oleic Acid 3-Chloro-2-hydroxypropyl-d5 Ester, 3-Chloro-1,2-propanediol-d5 1-Oleate, 3-Chloro-1,2-propanediol-d5 1-Monooleate
InChI formel InChI=1S/C21H39ClO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22/h9-10,20,23H,2-8,11-19H2,1H3/b10-9-/i18D2,19D2,20D
SMIL O=C(CCCCCCC/C=C\CCCCCCCC)OC([2H])([2H])C(O)([2H])C([2H])([2H])Cl
IUPAC navn 3-chloro-2-hydroxypropyl-1,1,2,3,3-d5 oleate
Molekylvægt (g/mol) 380.02
Formel vægt 379.29