Få mere at vide
rac-1-Linoleoyl-3-chloropropanediol, TRC
Artikelnummer. 30483963
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483963 50 mg
30476958 10 mg
30474117 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30483963 Leverandør Toronto Research Chemicals Leverandørnr. TRCL46780050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 74875-98-2
Molekylær formel C21 H37 Cl O3
Molekylvægt (g/mol) 372.97
Synonym 9,12-Octadecadienoic acid (Z,Z)-, 3-chloro-2-hydroxypropyl ester, 3-Chloro-1,2-propanediol 1-linoleate, Rac-1-linoleoyl-3-chloropropane-1,2-diol
IUPAC navn (3-chloro-2-hydroxypropyl) (9Z,12Z)-octadeca-9,12-dienoate
SMIL CCCCC\C=C/C\C=C/CCCCCCCC(=O)OCC(O)CCl
Tekniske data
Kemisk navn eller materiale rac-1-Linoleoyl-3-chloropropanediol
Mængde 50 mg
InChI formel InChI=1S/C21H37ClO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22/h6-7,9-10,20,23H,2-5,8,11-19H2,1H3/b7-6-,10-9-
Formel vægt 372.2431