missing translation for 'onlineSavingsMsg'
Få mere at vide

rac-1-Linoleoyl-3-chloropropanediol, TRC

Artikelnummer. 30476958
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476958 10 mg
30474117 25 mg
30483963 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30476958 Leverandør Toronto Research Chemicals Leverandørnr. TRCL46780010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74875-98-2
Molekylær formel C21 H37 Cl O3
Molekylvægt (g/mol) 372.97
Synonym 9,12-Octadecadienoic acid (Z,Z)-, 3-chloro-2-hydroxypropyl ester, 3-Chloro-1,2-propanediol 1-linoleate, Rac-1-linoleoyl-3-chloropropane-1,2-diol
IUPAC navn (3-chloro-2-hydroxypropyl) (9Z,12Z)-octadeca-9,12-dienoate
SMIL CCCCC\C=C/C\C=C/CCCCCCCC(=O)OCC(O)CCl

Specifications

Kemisk navn eller materiale rac-1-Linoleoyl-3-chloropropanediol
Mængde 10 mg
InChI formel InChI=1S/C21H37ClO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22/h6-7,9-10,20,23H,2-5,8,11-19H2,1H3/b7-6-,10-9-
Formel vægt 372.2431
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.